Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat IgG Antibody (GT175), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250413AF594
Description
Goat IgG Monoclonal specifically detects Goat IgG in Goat samples. It is validated for Immunohistochemistry.Specifications
Goat IgG | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
Protein A purified | |
Primary | |
Store at 4°C in the dark. |
Immunohistochemistry | |
GT175 | |
Immunohistochemistry | |
Immunoglobulin G | |
Goat immunoglobulins | |
0.1 mL | |
Goat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction