Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat IgG VisUCyte HRP Polymer, R&D Systems™

Donkey Polyclonal Antibody has been used in 2 publications
$245.00 - $2360.00
Specifications
Antigen | IgG |
---|---|
Dilution | Immunohistochemistry |
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | HRP Polymer |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
VC004025
![]() |
R&D Systems
VC004025 |
25 mL |
Each for $245.00
|
|
|||||
VC004050
![]() |
R&D Systems
VC004050 |
50 mL (2 x 25 mL) |
Each for $456.00
|
|
|||||
VC004125
![]() |
R&D Systems
VC004125 |
125 mL |
Each for $1,218.00
|
|
|||||
VC004250
![]() |
R&D Systems
VC004250 |
250 mL (2 x 125 mL) |
Each for $2,360.00
|
|
|||||
Description
IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Immunohistochemistry.Specifications
IgG | |
Immunohistochemistry | |
HRP Polymer | |
RUO | |
Immunoglobulin G, ImmunoglobulinG | |
Goat IgG | |
Secondary | |
6 months from date of receipt, 2 to 8 degreesC as supplied. |
Immunohistochemistry | |
Polyclonal | |
Donkey | |
Supplied as a solution in PBS with preservative. | |
3500 | |
IgG | |
Affinity Purified | |
Detects heavy ( gamma ) chains on goat IgG and light chains on all goat immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin goat serum proteins or serum proteins from human, mouse or rat, based on IEP. VisUCyte™ HRP Polymer provides high sensitivity with low background in IHC. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title