Learn More
Description
This DuoSet ELISA is intended for the development of sandwich ELISAs to measure natural and recombinant goat IL-6. It features optimized capture and detection antibody pairings to save development time.
- Assay Type: Solid Phase Sandwich ELISA
- Reactivity: Goat
- Assay Range: 31.3 to 2000 pg/mL
- Sample Type: Cell culture supernates, serum, and plasma
- Assay Format: 96-well strip plate
- Detection Method: Colorimetric ELISA - 450nm (TMB)
- Conjugate: Biotin
- Label: HRP
Specifications
Specifications
| Product Type | Solid Phase Sandwich ELISA |
| Quantity | 1 Kit (for 15 Plates) |
| Species | Goat |
| Assay Range | 31.30 to 2000 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3569 |
| Product Line | R&D Systems |
| Synonym | B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Research Area | Cytokines |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
