Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GOAT/MBOAT4 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP226066
Description
GOAT/MBOAT4 Polyclonal specifically detects GOAT/MBOAT4 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
GOAT/MBOAT4 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
MBOAT4 | |
Peptide with sequence C-QSPGEEGYVPDAD corresponding to internal region according to NP_001094386.1. | |
0.1 mg | |
Primary | |
Human | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1:100 - 1:2000, Peptide ELISA Detection limit 1:2000 | |
EC 2.3.1.-, ghrelin O-acyltransferase, GOATFKSG89, membrane bound O-acyltransferase domain containing 4, Membrane-bound O-acyltransferase domain-containing protein 4, OACT4, O-acyltransferase (membrane bound) domain containing 4, O-acyltransferase domain-containing protein 4 | |
Goat | |
Affinity Purified | |
RUO | |
619373 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction