Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Goat Rotavirus Antigen ELISA Kit (Colorimetric)
SDP

Catalog No. NB601311KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB601311KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB601311KT Supplier Novus Biologicals™ Supplier No. NBP2601311KIT

Direct ELISA Kit for detection of goat rotavirus antigen.

The Goat Rotavirus Antigen ELISA Kit (Colorimetric) measures Goat Rotavirus Antigen in feces.

Specifications

Assay Range Qualitative
Cross Reactivity No significant cross-reactivity or interference between goat rotavirus antigen and analogues was observed
Product Type ELISA Kit (Colorimetric)
Sample Type Fecal Extract
Target Species Goat
For Use With (Application) ELISA
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Cell Biology
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Target Rotavirus Antigen
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.