Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Goat Thyroxine ELISA Kit (Colorimetric)
SDP

Catalog No. NB601571KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB601571KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB601571KT Supplier Novus Biologicals™ Supplier No. NBP2601571KIT
Add to Cart
Add to Cart

Colorimetric ELISA Kit for study of thyroid function.

The Goat Thyroxine ELISA Kit (Colorimetric) measures goat Thyroxine in serum, plasma, tissue homogenates.

Specifications

Assay Range 20 to 320ng/mL
Assay Sensitivity 20ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum, Tissue Homogenates
Specificity For detection of goat T4
Target Species Goat
For Use With (Application) ELISA
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Signal Transduction
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Synonym T4
Target Thyroxine
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.