Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GODZ Polyclonal Antibody, Biotin, Bioss
SDP

Catalog No. 502281414
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1414 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1414 Supplier Bioss Supplier No. BS24751RBIOTIN
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GODZ Polyclonal antibody specifically detects GODZ in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen GODZ
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene ZDHHC3
Gene Accession No. Q8R173, Q9NYG2
Gene Alias 1110020O22Rik; 1810006O10Rik; 2210017C02Rik; DHHC1 protein; DHHC-3; GABA-A receptor-associated membrane protein 1; Godz; Golgi apparatus-specific protein with DHHC zinc finger domain; golgi-specific DHHC Zinc Finger Protein; Gramp1; HSD49; Palmitoyltransferase ZDHHC3; Protein DHHC1; Zdhhc3; Zfp373; Zinc finger DHHC domain-containing protein 3; zinc finger DHHC-type containing 3; zinc finger protein 373; zinc finger, DHHC domain containing 3; zinc finger, DHHC-type containing 3; ZNF373
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GODZ
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 301081, 51304, 69035
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.