Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold(III) chloride, Au 64.4% min, Alfa Aesar

Manufacturer:  Alfa Aesar 1216303

Quantity 1g

 View more versions of this product

Catalog No. AA1216303


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

13453-07-1
AuCl3
MFCD00014172
Crystalline
303.33
Gold(III) chloride

Specifications

13453-07-1
AuCl3
303.33
3.9g/cm3
Red
Gold(III) chloride
Crystalline
Red crystalline
MFCD00014172
Odorless
1g

Safety and Handling

SDS & Safety Documents

H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 8, Packing Group: III

EINECSNumber : 236-623-1

storageNote1 : Hygroscopic

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit