Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold(III) chloride trihydrate, Honeywell Fluka™

Catalog No. p-7072297 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 g
25 g

ACS Reagent,≥49.0% Au basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16961-25-4
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
MDL Number MFCD00149904
InChI Key VDLSFRRYNGEBEJ-UHFFFAOYSA-K
IUPAC Name trichlorogold hydrochloride
SMILES Cl.Cl[Au](Cl)Cl

Specifications

Quantity 1 g
Linear Formula HAuCl4 · 3H2O
UN Number UN3260
Formula Weight 393.83g/mol
Percent Purity ≥49% (Au basis)
Chemical Name or Material Gold(III) chloride trihydrate
Hazard Statement P280-P305 + P351 + P338-P310
RTECSNumber MD5428000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.