Learn More
Gold(III) oxide, Premion™, 99.99% (metals basis), Au 88.6% min
Description
It is used in aerospace and electrochemical applications such as fuel cells in which they exhibit ionic conductivity. It acts as strong reducing agent in redox reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 150°C (decomposition) |
| Color | Brown |
| Physical Form | Powder |
| Quantity | 1 g |
| Merck Index | 14,4524 |
| Solubility Information | Insoluble in water; Soluble in HCl,HNO3and in NaCN solution. |
| Formula Weight | 441.93 |
| Percent Purity | 99.99% |
| Grade | Premion |
| Concentration or Composition (by Analyte or Components) | Au 88.6% min. |
| Show More |
Safety and Handling
| Hazard Category | H315-H319 |
| Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
| EINECSNumber | 215-122-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.