Learn More
Gold Palladium wire, 0.2mm (0.008in) dia, annealed, 99.9% (metals basis)
Description
Gold Palladium wire is used as a microelectrode device, which exhibits excellent responses to hydrogen in concentrations as low as 0.5% by volume. It is also involved in the enhanced sensitivity for the detection of hydrogen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Gold Palladium wire |
| Name Note | 0.2mm (0.008 in.) dia., Annealed, Temper: softest 0.154g/ft |
| Avg. Mol. Wt. or Mol. Wt. Range | Au:Pd; 60:40 wt% |
| Quantity | 1 m |
| Form | Wire |
| Assay Percent Notes | (metals basis) |
| MDL Number | MFCD00003436 |
| Solubility Information | Insoluble in water. |
| Percent Purity | 99.9% |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.