Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar Gold Palladium wire, 0.2mm (0.008 in.) dia., annealed, 99.9% (metals basis)
Manufacturer: Alfa Aesar 41206BU
Specifications
Au:Pd; 60:40 wt% | |
25cm | |
(metals basis) | |
Insoluble in water. | |
99.9% |
Gold Palladium wire | |
0.2mm (0.008 in.) dia., Annealed, Temper: softest 0.154g/ft | |
MFCD00003436 | |
Wire, Temper: soft est .154g/ft |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title