Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Gold Platinum Palladium powder, APS <5 micron, 99.9% (metal s basis)

$246.00 - $886.00

Specifications

Chemical Name or Material Gold Platinum Palladium powder
MDL Number MFCD00192403
UN Number UN3089
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA8972603 View Documents Alfa Aesar™
8972603
1g Each for $246.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
AA8972606 View Documents Alfa Aesar™
8972606
5g Each for $886.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

Gold Platinum Palladium powder
MFCD00192403
UN3089
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit