Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold Tin eutectic foil, 0.1mm (0.004 in.) thick x 43.8mm (1.72in) wide, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7050253
Click to view available options
Quantity:
10 cm

Gold Tin eutectic foil is used in electronic manufacturing. It can be made into dimensions much smaller than soft solders, thereby it is utilized in applications which require the highest level of precision.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gold Tin eutectic foil
Name Note 0.1mm (0.004 in.) thick x 43.8mm (1.72 in.) wide
Avg. Mol. Wt. or Mol. Wt. Range Au:Sn; 80:20 wt%
Quantity 10 cm
Weight ≈6.37g/10cm
Form Foil
Assay Percent Notes (metals basis)
MDL Number MFCD02091709
Solubility Information Insoluble in water.
Percent Purity 99.99%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.