Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Golden Guides
SCI_ED

Catalog No. S51432
Click to view available options
Product Type:
Endangered Animals
Pond Life
Whales, Dolphins and Other Marine Mammals

A student and teacher favorite.

  • Golden Guides have been a science classroom staple for years
  • Full of colorful illustrations and informative easy-to-read text
  • Paperback, 4 x 6 in.

Specifications

Author George K. Reid
Format Paperback Book
Product Type Pond Life
For Use With (Application) Describe and illustrates, in full color, the plants and animals that live in or near ponds, lakes, streams and wetlands
Subject Biology
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.