Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLGA1 Mouse anti-Human, Clone: CDF4, Abnova™

Catalog No. 89035724 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-724 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-724 Supplier Abnova Corporation Supplier No. MAB14066

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human GOLGA1.

  • High specificity
  • High reactivity

Specifications

Antigen GOLGA1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone CDF4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human GOLGA1.
Dilution Immunocytochemistry (1 ug/mL) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Formulation Lyophilized.
Gene Accession No. Q92805
Gene Alias MGC33154/golgin-97
Gene Symbols GOLGA1
Host Species Mouse
Immunogen Human GOLGA1.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2800
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.