Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLGA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518419
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518419 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518419 Supplier Invitrogen™ Supplier No. PA518419
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

The protein encoded by this gene is a member of kinesin-like protein family. Proteins of this family are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. This protein is important for anaphase chromosome segregation and may be required to coordinate the onset of sister centromere separation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLGA3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Golga3
Gene Accession No. Q08378
Gene Alias 5330413L04; 5430416E01Rik; AI449376; AW490576; G1-499-14; GCP170; GOLGA3; golgi autoantigen, golgin subfamily a, 3; golgi complex-associated protein of 170 kDa; Golgi membrane associated protein; Golgi peripheral membrane protein; golgin A3; Golgin subfamily A member 3; Golgin-160; golgin-165; HGNC:4426; male enhanced antigen-2; male-enhanced antigen 2; Mea2; MEA-2; Mea2/Golga3; repro27; SY2/SY10 protein
Gene Symbols Golga3
Host Species Goat
Immunogen Synthetic peptide sequence (HSQSRASKEGPGE) corresponding to the C-terminus amino acids of GOLGA3 (aa 1486-1498).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2802
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.