Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLGA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576521
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576521 Supplier Invitrogen™ Supplier No. PA576521
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes one of the golgins, a family of proteins localized to the Golgi. This protein has been postulated to play a role in Rab6-regulated membrane-tethering events in the Golgi apparatus. Alternatively spliced transcript variants encoding different isoforms have been identified in this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLGA4
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene GOLGA4
Gene Accession No. Q13439, Q5U4E6, Q91VW5
Gene Alias 256 kDa golgin; 72.1 protein; AI225887; AU019508; centrosome-related protein F46; CRPF46; EMBL:AAH85124.1, ECO:0000312; GCP2; GOLG; Golga4; golga4 {ECO:0000312; golgi autoantigen, golgin subfamily a, 4; golgin A4; Golgin subfamily A member 4; golgin subfamily A member 4 {ECO:0000250; golgin-240; golgin-245; MU-RMS-40.18; Olp-1; p230; protein 72.1; RGD:1591925}; tGolgin-1; trans-Golgi p230; UniProtKB:Q13439}
Gene Symbols GOLGA4
Host Species Rabbit
Immunogen Recombinant fusion protein of human GOLGA4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2803, 501069, 54214
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.