Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GOLGA5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23900725UL
Description
GOLGA5 Polyclonal specifically detects GOLGA5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GOLGA5 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Q8TBA6 | |
GOLGA5 | |
This antibody was developed against a recombinant protein corresponding to amino acids: QEFHYIEEDLYRTKNTLQSRIKDRDEEIQKLRNQLTNKTLSNSSQSELENRLHQLTETLIQKQTMLESLSTEKNSLVFQLERLEQQMNS | |
25 μL | |
Golgi Apparatus Markers | |
9950 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Cell proliferation-inducing gene 31 protein, golgi autoantigen, golgin subfamily a, 5, golgin A5, Golgin subfamily A member 5, Golgin-84, GOLIM5, Protein Ret-II, PTC5, RET-fused gene 5 protein, ret-II, RETII, rfg5, RFG5golgi integral membrane protein 5 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction