Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLGA5 Monoclonal Antibody (GT2726)
GREENER_CHOICE

Catalog No. PIMA531564
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531564 Supplier Invitrogen™ Supplier No. MA531564
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (89%), Rat (88%), Rhesus Monkey (98%), Bovine (90%). Positive Control: A549. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes a member of the golgin family of proteins, whose members localize to the Golgi. This protein is a coiled-coil membrane protein that has been postulated to play a role in vesicle tethering and docking. Translocations involving this gene and the ret proto-oncogene have been found in tumor tissues; the chimeric sequences have been designated RET-II and PTC5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLGA5
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT2726
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GOLGA5
Gene Accession No. Q8TBA6
Gene Alias Cell proliferation-inducing gene 31 protein; GOLGA5; golgi autoantigen, golgin subfamily a, 5; golgi integral membrane protein 5; golgin A5; Golgin subfamily A member 5; Golgin-84; GOLIM5; PIG31; Protein Ret-II; Protein Sumiko; RET-fused gene 5; RET-fused gene 5 protein; RETII; ret-II; RFG5
Gene Symbols GOLGA5
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human GOLGA5. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9950
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.