Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLGA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590689
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590689 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590689 Supplier Invitrogen™ Supplier No. PA590689
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSWFVDLAGK AEDLLNRVDQ GAATALSRKD NASNIYSKNT DYTELHQQNT DLIYQTGPKS TYISSAADNI RNQKATILAG TANVKVGSRT PVEASHPVEN ASVPRPSSHF VRRKKSEPDD ELLFDFLNSS QKEPTGRVEI RKEKGKTPVF QSSQTSSVSS VNPSVTTIKT IEENSFGSQT HEAASNSDSS HEGQEESSKE NVSSNAACPD HTPTPNDDGK SHELSNLRLE NQLLRNEVQS LNQEMASLLQ RSKETQEELN KARARVEKWN ADHSKSDRMT RGLRAQVDDL TEAVAAKDSQ; Positive Samples: LO2, Jurkat, HeLa, Mouse brain, Mouse heart, Mouse kidney, Mouse liver, Rat testis; Cellular Location: Golgi apparatus membrane, Single-pass type IV membrane protein.

The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes a member of the golgin family of proteins, whose members localize to the Golgi. This protein is a coiled-coil membrane protein that has been postulated to play a role in vesicle tethering and docking. Translocations involving this gene and the ret proto-oncogene have been found in tumor tissues; the chimeric sequences have been designated RET-II and PTC5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLGA5
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GOLGA5
Gene Accession No. Q3ZU82, Q8TBA6, Q9QYE6
Gene Alias Cell proliferation-inducing gene 31 protein; GOLGA5; golgi autoantigen, golgin subfamily a, 5; golgi integral membrane protein 5; golgin A5; Golgin subfamily A member 5; Golgin-84; GOLIM5; PIG31; Protein Ret-II; Protein Sumiko; RET-fused gene 5; RET-fused gene 5 protein; RETII; ret-II; RFG5
Gene Symbols GOLGA5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human GOLGA5 (NP_0051043).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27277, 299258, 9950
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.