Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLGA6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562132
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562132 Supplier Invitrogen™ Supplier No. PA562132
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALCAVSTQQQ EEDRSSSCRE AVLHRRLQQT IKERAL Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 33%.

This gene is found in a large, low copy repeat sequence or duplicon that is found in multiple copies, which are greater than 90% similar, on chromosome 15. Duplicons are associated with deletions, inversions and other chromosomal rearrangements that underlie genomic disease. This gene is a member of the golgin gene family, whose protein products localize to the Golgi apparatus. The majority of the related gene copies are thought to be transcribed pseudogenes. It is not known whether this gene is a pseudogene or if it encodes a golgin protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLGA6B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GOLGA6B
Gene Accession No. A6NDN3
Gene Alias GOLGA; GOLGA6B; GOLGA6D; golgi autoantigen, golgin subfamily a, 6B; golgin A6 family member B; golgin A6 family, member B; golgin subfamily A member 6B; golgin-like protein; putative golgin subfamily A member 6B; Putative golgin subfamily A member 6D
Gene Symbols GOLGA6B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GOLGA6B. Recombinant protein control fragment (Product #RP-99520).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55889
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.