Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GOLGA6B Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142178
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:1,000.
This gene is found in a large, low copy repeat sequence or duplicon that is found in multiple copies, which are greater than 90% similar, on chromosome 15. Duplicons are associated with deletions, inversions and other chromosomal rearrangements that underlie genomic disease. This gene is a member of the golgin gene family, whose protein products localize to the Golgi apparatus. The majority of the related gene copies are thought to be transcribed pseudogenes. It is not known whether this gene is a pseudogene or if it encodes a golgin protein.
Specifications
GOLGA6B | |
Polyclonal | |
Unconjugated | |
GOLGA6B | |
GOLGA; GOLGA6B; GOLGA6D; golgi autoantigen, golgin subfamily a, 6B; golgin A6 family member B; golgin A6 family, member B; golgin subfamily A member 6B; golgin-like protein; putative golgin subfamily A member 6B; Putative golgin subfamily A member 6D | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
55889 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
A6NDN3 | |
GOLGA6B | |
Peptide with sequence C-DLPKEKADGTEQ. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction