Learn More
Invitrogen™ GOLGA6L2 Polyclonal Antibody

Description
GOLGA6L2 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | GOLGA6L2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | GOLGA6L2 |
| Gene Accession No. | Q8N9W4 |
| Gene Alias | cancer/testis antigen 105; CT105; GOLGA6L2; golgi autoantigen, golgin subfamily a, 6-like 2; golgin A6 family-like 2; golgin subfamily A member 6-like protein 2 |
| Gene Symbols | GOLGA6L2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.