Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLGA6L6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571116
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571116 Supplier Invitrogen™ Supplier No. PA571116
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

GOLGA6L6 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLGA6L6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GOLGA6L6
Gene Accession No. A8MZA4
Gene Alias GOLGA6L6; golgi autoantigen, golgin subfamily a, 6-like 6; golgin A6 family-like 6; golgin subfamily A member 6-like protein 6; putative golgin subfamily A member 6-like protein 6
Gene Symbols GOLGA6L6
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human GOLGA6L6.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727832
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.