Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLGA7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119708
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119708 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119708 Supplier Invitrogen™ Supplier No. PA5119708
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Human stomach tissue lysate, A431 cell lysate, human breast cancer tissue, human kidney tissue, mouse kidney tissue, A431. Subcellular Location: Golgi apparatus, Membrane.

GOLGA7 may be involved in protein transport from Golgi to cell surface. The ZDHHC9-GOLGA7 complex is a palmitoyltransferase specific for HRAS and NRAS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLGA7
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene GOLGA7
Gene Accession No. Q7Z5G4, Q91W53
Gene Alias AB041568; C130038N16Rik; GCP16; GOLGA3AP1; GOLGA7; GOLGA7A; golgi autoantigen, golgin subfamily a, 7; golgi complex-associated protein of 16 kDa; Golgi complex-associated protein of 16kDa; golgin A7; Golgin subfamily A member 7; Golgin subfamily A member 7; golgin subfamily A member 7; golgin subfamily A member 7-like; HDCKB03P; HSPC041; hypothetical protein LOC415226; MNCb-1213; R75586; wu:fc23b09; zgc:86753
Gene Symbols GOLGA7
Host Species Rabbit
Immunogen Recombinant protein within Human GOLGA7 aa 1-137.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51125, 57437
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.