Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLGA7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107136
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107136 Supplier Thermo Scientific Supplier No. PA5107136
Add to Cart
Add to Cart

GOLGA7 Polyclonal Antibody

GOLGA7 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLGA7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene GOLGA7
Gene Accession No. Q7Z5G4
Gene Alias AB041568; C130038N16Rik; GCP16; GOLGA3AP1; GOLGA7; GOLGA7A; golgi autoantigen, golgin subfamily a, 7; golgi complex-associated protein of 16 kDa; Golgi complex-associated protein of 16kDa; golgin A7; Golgin subfamily A member 7; Golgin subfamily A member 7; golgin subfamily A member 7; golgin subfamily A member 7-like; HDCKB03P; HSPC041; hypothetical protein LOC415226; MNCb-1213; R75586; wu:fc23b09; zgc:86753
Gene Symbols GOLGA7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human GOLGA7 (aa 71-121).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51125
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.