Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLGA7 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982906
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2906 Supplier Bioss Supplier No. BS13484R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GOLGA7 may be involved in protein transport from Golgi to cell surface. The ZDHHC9-GOLGA7 complex is a palmitoyltransferase specific for HRAS and NRAS.

Specifications

Antigen GOLGA7
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GOLGA7
Gene Accession No. Q7Z5G4
Gene Alias AB041568; BOS_23907; C130038N16Rik; GCP16; GOLGA3AP1; Golga7; GOLGA7A; golgi autoantigen, golgin subfamily a, 7; Golgi complex-associated protein of 16 kDa; Golgi complex-associated protein of 16kDa; golgin A7; Golgin subfamily A member 7; Golgin subfamily A member 7; golgin subfamily A member 7; golgin subfamily A member 7-like; HDCKB03P; HSPC041; hypothetical protein LOC415226; LOC100156903; MNCb-1213; R75586; wu:fc23b09; zgc:86753
Gene Symbols GOLGA7
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GOLGA7.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51125
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.