Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLGA8A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535036
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535036 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535036 Supplier Invitrogen™ Supplier No. PA535036
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GOLGA8A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

May be involved in maintaining Golgi structure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLGA8A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GOLGA8A
Gene Accession No. A7E2F4
Gene Alias 88 kDa Golgi matrix protein; 88-kDa golgi protein; GM88; GM88 autoantigen; GOLGA8A; golgi autoantigen, golgin subfamily a, 8A; golgin A8 family member A; golgin A8 family, member A; golgin subfamily A member 8A; golgin-67; KIAA0855
Gene Symbols GOLGA8A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 129-157 amino acids from the N-terminal region of human GOG8A.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23015
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.