Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLGA8B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549042
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549042 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549042 Supplier Invitrogen™ Supplier No. PA549042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GOLGA8B Polyclonal Antibody for Western Blot

May be involved in maintaining Golgi structure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLGA8B
Applications Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GOLGA8B
Gene Accession No. A8MQT2
Gene Alias GOLGA5; GOLGA8B; golgi autoantigen, golgin subfamily a, 8B; golgin A8 family member B; golgin A8 family, member B; golgin subfamily A member 8B; Golgin-67
Gene Symbols GOLGA8B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 18-44 amino acids from the N-terminal region of human GOLGA8B.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440270
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.