Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLGA8B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989802
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9802 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9802 Supplier Bioss Supplier No. BS16257R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be involved in maintaining Golgi structure.

Specifications

Antigen GOLGA8B
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene GOLGA8B
Gene Accession No. A8MQT2
Gene Alias GOLGA5; GOLGA8B; golgi autoantigen, golgin subfamily a, 8B; golgin A8 family member B; golgin A8 family, member B; Golgin subfamily A member 8B; Golgin-67
Gene Symbols GOLGA8B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GOLGA8B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440270
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.