Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLGB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100542 Supplier Invitrogen™ Supplier No. PA5100542
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Giantin.

Golgin subfamily B member 1 (GOLGB1) is a single pass type I membrane protein located in the golgi apparatus membrane. GOLGB1 may participate in forming intercisternal cross-bridges of the Golgi complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLGB1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GOLGB1
Gene Accession No. Q14789
Gene Alias 372 kDa Golgi complex-associated protein; 4930428L02Rik; 628101; 6330407A06Rik; AU042952; C130074L01Rik; F730017E11Rik; GCP; GCP372; Giantin; Gm6840; GOLGB1; golgi autoantigen, golgin subfamily b, macrogolgin (with transmembrane signal), 1; golgi autoantigen, golgin subfamily b, macrogolgin 1; golgi integral membrane protein 1; golgi-associated protein GCP360; golgin B1; golgin B1, golgi integral membrane protein; golgin subfamily B member 1; Golgin subfamily B member 1; LOW QUALITY PROTEIN: golgin subfamily B member 1; golgin subfamily b member 1-like protein; GOLIM1; LOW QUALITY PROTEIN: golgin subfamily B member 1; macrogolgin; mKIAA4151
Gene Symbols GOLGB1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GOLGB1(Accession Q14789), corresponding to amino acid residues E1787-N1837.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2804
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.