Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GOLGB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552841
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552841 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552841 Supplier Invitrogen™ Supplier No. PA552841
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLSGLANVVL HELSGDDDTD QNMRAPLDPE LHQESDMEFN NTTQEDVQER LAYAEQLVVE LKDIIRQKDV QLQQKDEALQ EERKAADNKI KKLKLHAKAK LTSLNKYIEE MKAQGGTVLP TEPQSEEQLS KHD Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 67%.

Golgin subfamily B member 1 (GOLGB1) is a single pass type I membrane protein located in the golgi apparatus membrane. GOLGB1 may participate in forming intercisternal cross-bridges of the Golgi complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLGB1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GOLGB1
Gene Accession No. Q14789
Gene Alias 372 kDa Golgi complex-associated protein; 4930428L02Rik; 628101; 6330407A06Rik; AU042952; C130074L01Rik; F730017E11Rik; GCP; GCP372; Giantin; Gm6840; GOLGB1; golgi autoantigen, golgin subfamily b, macrogolgin (with transmembrane signal), 1; golgi autoantigen, golgin subfamily b, macrogolgin 1; golgi integral membrane protein 1; golgi-associated protein GCP360; golgin B1; golgin B1, golgi integral membrane protein; golgin subfamily B member 1; Golgin subfamily B member 1; LOW QUALITY PROTEIN: golgin subfamily B member 1; golgin subfamily b member 1-like protein; GOLIM1; LOW QUALITY PROTEIN: golgin subfamily B member 1; macrogolgin; mKIAA4151
Gene Symbols GOLGB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GOLGB1. Recombinant protein control fragment (Product #RP-90447).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2804
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.