Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLGB1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501566721
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6721 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6721 Supplier Bethyl Laboratories, Inc Supplier No. A304034AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

GOLGB1 may participate in forming intercisternal cross-bridges of the Golgi complex.

Specifications

Antigen GOLGB1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene GOLGB1
Gene Accession No. Q14789
Gene Alias 372 kDa Golgi complex-associated protein, GCP, GCP372, giantin, golgi autoantigen, golgin subfamily b, macrogolgin (with transmembrane signal), 1, golgi integral membrane protein 1, golgin B1, golgi integral membrane protein, GOLIM1, macrogolgin
Gene Symbols GOLGB1
Host Species Rabbit
Immunogen Between 950 and 981
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2804
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.