Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Golgi Complex Antibody (371-4), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234534F
Description
Golgi Complex Monoclonal specifically detects Golgi Complex in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Golgi Complex | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
SU-DHL-1 large cell lymphoma cells. | |
0.1 mL | |
Primary | |
Human, Mouse (Negative), Rat (Negative) | |
IgG1 κ |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
371-4 | |
Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Mouse | |
Protein A or G purified | |
RUO | |
This monoclonal antibody recognizes an antigen associated with the Golgi complex in human cells only. It can be used to stain the Golgi complex in cell or tissue preparations and can be used as a Golgi marker in subcellular fractions. It produces a diffuse staining pattern of the Golgi zone in normal and malignant cells. This monoclonal antibody is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells. The Golgi apparatus is an organelle present in all eukaryotic cells that forms a part of the endomembrane system. The primary function of the Golgi apparatus is to process and package macromolecules synthesized by the cell for exocytosis or use within the cell. The Golgi is made up of a stack of flattened, membrane-bound sacs known as cisternae, with three functional regions: the cis face, medial region and trans face. Each region consists of various enzymes that selectively modify the macromolecules passing though them, depending on where they are destined to reside. Several spherical vesicles that have budded off of the Golgi are present surrounding the main cisternae. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction