Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Golgi Complex Antibody (371-4), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234534MFV450
Description
Golgi Complex Monoclonal antibody specifically detects Golgi Complex in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Flow CytometrySpecifications
Golgi Complex | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Human, Mouse (Negative), Rat (Negative) | |
Purified |
Flow Cytometry | |
371-4 | |
Flow Cytometry | |
AI593353;AW537898;CFR;CFR1;CFR-1;ESL1;ESL-1;GLG1;MG160;MG-160;Selel | |
SU-DHL-1 large cell lymphoma cells. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction