Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Golgi Complex Antibody (AE-6), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311427H
Description
Golgi Complex Monoclonal antibody specifically detects Golgi Complex in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Golgi Complex | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Human, Mouse (Negative), Rat (Negative) | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
AE-6 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
AI593353;AW537898;CFR;CFR1;CFR-1;ESL1;ESL-1;GLG1;MG160;MG-160;Selel | |
SU-DHL-1 large cell lymphoma cells | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction