Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Golgi Complex Antibody (GLG1/2829R), Alexa Fluor™ 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308396AF405
Description
Golgi Complex Monoclonal antibody specifically detects Golgi Complex in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Golgi Complex | |
Monoclonal | |
Alexa Fluor 405 | |
AI593353;AW537898;CFR;CFR1;CFR-1;ESL1;ESL-1;GLG1;MG160;MG-160;Selel | |
Recombinant full-length human Golgi Complex protein (Uniprot: Q92896) | |
100 μg | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
GLG1/2829R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Human, Mouse (Negative), Rat (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction