Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Golgi Complex Rabbit anti-Human, Clone: GLG1/2829R, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30727820UG
Description
Golgi Complex Monoclonal specifically detects Golgi Complex in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Golgi Complex | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Rabbit | |
Protein A or G purified | |
RUO | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
GLG1/2829R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
SU-DHL-1 large cell lymphoma cells. | |
20 μg | |
Primary | |
Store at 4°C. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction