Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Golgi phosphoprotein 6 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982914
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2914 Supplier Bioss Supplier No. BS13492R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GRASP55 is a PDZ containing protein involved in the stacking of golgi cisternea in a cell-free system.

Specifications

Antigen Golgi phosphoprotein 6
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GORASP2
Gene Accession No. Q9H8Y8
Gene Alias 0610011A07Rik; 2410043M02Rik; 5730520M13Rik; 9430094F20Rik; AW552058; Golgi phosphoprotein 6; golgi reassembly stacking protein 2; golgi reassembly stacking protein 2, 55kDa; Golgi reassembly-stacking protein 2; Golgi reassembly-stacking protein of 55 kDa; GOLPH2; GOLPH6; Gorasp2; GRASP55; GRS2; p59
Gene Symbols GORASP2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Golgi phosphoprotein 6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26003
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.