Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Golgi protein 58k Monoclonal Antibody (58K 9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA546614
This item is not returnable.
View return policy
Description
Golgi protein 58k Monoclonal Antibody for Western Blot, ICC/IF, Flow
Golgi Protein, 58K is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool. Mutations in this gene are associated with glutamate formiminotransferase deficiency. Alternatively spliced transcript variants have been found for this gene.
Specifications
Golgi protein 58k | |
Monoclonal | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O88618, O95954, Q91XD4, Q9YH58 | |
Ftcd | |
Full length native protein (purified). | |
100 μg | |
Primary | |
Human, Mouse, Rat, Canine, Bovine, Chicken | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot, Immunocytochemistry | |
58K 9 | |
Unconjugated | |
Ftcd | |
58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1 | |
Mouse | |
Protein A | |
RUO | |
10841, 14317, 395726, 535816, 607685, 89833 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction