Learn More
Invitrogen™ Golgi protein 58k Monoclonal Antibody (GT575)

Description
Keep as concentrated solution. Predicted reactivity: Pig (85%), Bovine (81%). Positive Control: HepG2, mouse liver. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Golgi protein 58k |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | GT575 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | Ftcd |
| Gene Accession No. | O95954, Q91XD4 |
| Gene Alias | 58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.