Learn More
Invitrogen™ Golgi protein 58k Polyclonal Antibody

Description
Immunogen sequence: MSQLVECVPN FSEGKNQEVI DAISGAITQT PGCVLLDVDA GPSTNRTVYT FVGPPECVVE GALNAARVAS RLIDMSRHQG EH Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 89%.
Specifications
Specifications
| Antigen | Golgi protein 58k |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Ftcd |
| Gene Accession No. | O95954 |
| Gene Alias | 58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1 |
| Gene Symbols | Ftcd |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.