Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Golgi protein 58k Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597218
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597218 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597218 Supplier Invitrogen™ Supplier No. PA597218
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DAEAFTAYLE AMRLPKNTPE EKDRRTAALQ EGLRRAVSVP LTLAETVASL WPALQELARC GNLACRSDLQ VAAKALEMGV FGAYFNVLIN LRDITDEAFK DQIHHRVSSL LQEAKTQAAL VLDCLETRQE; Positive Samples: LO2, Mouse liver, Mouse kidney, Rat liver, Rat kidney; Cellular Location: Cytoplasm, Golgi apparatus, centriole, centrosome, cytoskeleton, microtubule organizing center.

Golgi Protein, 58K is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool. Mutations in this gene are associated with glutamate formiminotransferase deficiency. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Golgi protein 58k
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ftcd
Gene Accession No. O88618, O95954, Q91XD4
Gene Alias 58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1
Gene Symbols Ftcd
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 412-541 of human FTCD (NP_0066481).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10841, 14317, 89833
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.