Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
golgi transport 1 homolog B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | golgi transport 1 homolog B |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
golgi transport 1 homolog B Polyclonal specifically detects golgi transport 1 homolog B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
golgi transport 1 homolog B | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
CGI-141, Golgi transport 1 homolog B, golgi transport 1 homolog B (S. cerevisiae), golgi transport 1B, GOT1, GOT1A, hGOT1a, Putative NF-kappa-B-activating protein 470, vesicle transport protein GOT1B, YMR292W | |
GOLT1B | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
51026 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RRVPVLGSLLNLPGIRSFVDKVGESN | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title