Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Golgicide A

Catalog No. 358410R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-841-0R 10 mg
35-845-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-841-0R Supplier Tocris Bioscience™ Supplier No. 3584/10
This item is not returnable. View return policy

Potent, selective and reversible inhibitor of GBF1 ArfGEF

Golgicide A is a potent, specific and reversible inhibitor of Golgi BFA resistance factor 1 (GBF1), an ArfGEF, that decreases Arf1 activation in vivo. Arrests secretion of soluble and membrane-associated proteins at the endoplasmic reticulum-Golgi intermediate compartment. Causes disassembly and dispersal of the Golgi and trans-Golgi network.

Specifications

Purity >98%
Quantity 10 mg
Product Type Golgicide A
Molecular Weight (g/mol) 284.3
Inhibitors ARF GTPase Inhibitors

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.