Learn More
Golgin-160 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303404AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae . Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes a member of the golgin family of proteins which are localized to the Golgi. Its encoded protein has been postulated to play a role in nuclear transport and Golgi apparatus localization. Several alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.Specifications
Golgin-160 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q08378 | |
GOLGA3 | |
Between 1448 and 1498 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GOLGA3 | |
GOLGA3; HGNC:4426; GCP170; MEA-2 ; Golgi autoantigen, golgin subfamily a, 3; Golgi complex-associated protein of 170 kD; Golgi membrane associated protein; Golgi peripheral membrane protein; SY2/SY10 protein; golgin-160; golgin-165; male enhanced antigen-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2802 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.