Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Golgin 160 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982902
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2902 Supplier Bioss Supplier No. BS13480R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a member of kinesin-like protein family. Proteins of this family are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. This protein is important for anaphase chromosome segregation and may be required to coordinate the onset of sister centromere separation.

Specifications

Antigen Golgin 160
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Golga3
Gene Accession No. Q08378
Gene Alias 5330413L04; 5430416E01Rik; AI449376; AW490576; G1-499-14; GCP170; GOLGA3; golgi autoantigen, golgin subfamily a, 3; Golgi complex-associated protein of 170 kDa; Golgi membrane associated protein; Golgi peripheral membrane protein; golgin A3; Golgin subfamily A member 3; Golgin-160; golgin-165; HGNC:4426; male enhanced antigen-2; Male-enhanced antigen 2; Mea2; MEA-2; Mea2/Golga3; repro27; SY2/SY10 protein
Gene Symbols Golga3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Golgin 160.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2802
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.