Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Golgin 97 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281753AF488
This item is not returnable.
View return policy
Description
Golgin 97 Polyclonal antibody specifically detects Golgin 97 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Golgin 97 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
gap junction protein, alpha 4, 37kD, golgi autoantigen, golgin subfamily a, 1, golgin A1, Golgin subfamily A member 1, Golgin-97, MGC33154 | |
Golgin 97 antibody was raised against a 17 amino acid peptide near the amino terminus of human Golgin 97. The immunogen is located within amino acids 30 - 80 of Golgin 97. Amino Acid Squence: DFASDGSSSREDLSSQL | |
0.1 mL | |
Golgi Apparatus Markers | |
2800 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction