Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Golgin 97 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$254.42 - $727.54
Specifications
| Antigen | Golgin 97 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Golgin 97 Polyclonal specifically detects Golgin 97 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Golgin 97 | |
| Polyclonal | |
| Rabbit | |
| Golgi Apparatus Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| gap junction protein, alpha 4, 37kD, golgi autoantigen, golgin subfamily a, 1, golgin A1, Golgin subfamily A member 1, Golgin-97, MGC33154 | |
| GOLGA1 | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q92805 | |
| 2800 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TGAESKITALEQKEQELQALIQQLSIDLQKVTAETQEKEDVITHLQEKVASLEKRLEQNLSGEEHLQELLK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title